Learn More
Sino Biological Recombinant Anti-TPO/Thrombopoietin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10381R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human THPO (rh THPO; Catalog#10381-H07H; NP_000451.1; Ser22-Gly353).
Catalog No. 50-172-5226