Learn More
Sino Biological Recombinant Human Contactin-1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10383H08H100
Encompass_Preferred
A DNA sequence encoding the amino acid residues (Met1-Ser993) of human CNTN1 (NP_001834.2) was expressed with the fused C-terminal polyhistidine tag.
Catalog No. 50-161-4808