Learn More
Sino Biological Anti-Thioredoxin/TRX Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10384RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Thioredoxin / TXN (rh Thioredoxin / TXN; Catalog#10384-HNAE; P10599; Met 1-Val 105). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5257