Learn More
Sino Biological Recombinant Human LIGHT/TNFSF14 Protein (His Tag)
Supplier:  Sino Biological 10386H07H100
Encompass_Preferred
A DNA sequence encoding the human TNFSF14 (NP_003798.2) extracellular domain (Asp 74-Val 240) was fused with a polyhistidine-tag at the N-terminus.
Catalog No. 50-161-4818