Learn More
Sino Biological Recombinant Human CD16a/FCGR3A/Fc gamma RIIIA Protein (F176, His Tag), HPLC-verified
Supplier:  Sino Biological 10389H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 208) of human FCGR3A (P08637-1, with Natural variant F176) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4827