Learn More
Sino Biological Anti-CD16a/FCGR3A/Fc gamma RIIIA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10389MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD16a (rhCD16a; Catalog#10389-H08H; P08637-1; Met1-Gln208,176F). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5263