Learn More
Sino Biological Anti-CD16a/FCGR3A/Fc gamma RIIIA Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10389MM41P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD16a (rh CD16a; Catalog#10389-H08C; P08637-1; Met1-Gln208,176F) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-5297