Learn More
Sino Biological Recombinant Anti-CD16/FCGR3/Fc gamma RIII Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10389R221F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD16a/Fc gamma RIIIa (rh CD16a/Fc gamma RIIIa; Catalog#10389-H08H; P08637-1; Met1-Gln208,176F) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-5301