Learn More
Sino Biological Anti-CD16a/FCGR3A/Fc gamma RIIIA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10389T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD16a/Fc gamma RIIIa (rh CD16a/Fc gamma RIIIa; Catalog#10389-H08H; P08637-1; Met1-Gln208,176F). CD16a/Fc gamma RIIIa specific IgG was purified by Human CD16a/Fc gamma RIIIa affinity chromatography.
Catalog No. 50-172-5308