Learn More
Sino Biological Anti-TIM-3/HAVCR2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10390MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TIMD3 (rh TIMD3; Catalog#10390-H08H; AAL65157.1; Met 1-Arg 200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5312