Learn More
Sino Biological Recombinant Anti-TIM-3/HAVCR2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10390R108100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TIM-3 / HAVCR2 (rh TIM-3 / HAVCR2 ; Catalog#10390-H08H; AAL65157.1; Met1-Arg200).
Catalog No. 50-172-5317