Learn More
Sino Biological Recombinant Human PGD Protein (His Tag)
Supplier:  Sino Biological 10393H07E100
Encompass_Preferred
A DNA sequence encoding the human PGD (P52209) (Met 1-Ala 483) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-4846