Learn More
Sino Biological Anti-IL-6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10395RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL6. Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5359