Learn More
Sino Biological Recombinant Human TIMP2 Protein
Supplier:  Sino Biological 10396HNAH20
Encompass_Preferred
A DNA sequence encoding the mature form of human TIMP2 (NP_003246.1) (Cys 27-Pro 220) was expressed and purified.
Catalog No. 50-161-4856