Learn More
Sino Biological Recombinant Human IL-20R alpha/IL-20RA Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10397H02H100
Encompass_Preferred
A DNA sequence encoding the human IL20RA (NP_055247.3) (Met1-Lys250) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4857