Learn More
Sino Biological Anti-IL-20R alpha/IL-20RA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10397MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL20RA isoform 1 (rh IL20RA; Catalog#10397-H08H; NP_055247.3; Met 1-Lys 250). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5378