Learn More
Sino Biological Recombinant Human IL-6R Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10398H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL6R (NP_000556.1) corresponding to amino acid (Met1-Pro365) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4861