Learn More
Sino Biological Anti-IL-6R Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10398MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL6 receptor extracellular domain (rhIL6R; Catalog#10398-H08H; NP_000556.1; Met 1-Pro 365). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5396