Learn More
Sino Biological Anti-IL-6R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10398RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL6 receptor extracellular domain (rhIL6R; Catalog#10398-H08H; NP_000556.1; Met 1-Pro 365). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5408