Learn More
Sino Biological Anti-IL-6R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10398T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-6R / CD126 (rh IL-6R / CD126; Catalog#10398-H08H; NP_000556.1; Met1-Pro365). IL-6R / CD126 specific IgG was purified by Human IL-6R / CD126 affinity chromatography.
Catalog No. 50-172-5414