Learn More
Sino Biological Recombinant Human CD4 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10400H08H100
Encompass_Preferred
A DNA sequence encoding the human CD4 (NP_000607.1) extracellular domain (Met 1-Trp 390) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4865