Learn More
Sino Biological Anti-CD4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10400RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD4 / Leu-3 (rh CD4 / Leu-3; Catalog#10400-H08H; NP_000607.1; Met 1-Trp 390). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5447