Learn More
Sino Biological Recombinant Human IL-4R Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10402H08H100
Encompass_Preferred
A DNA sequence encoding the human IL4R (NP_000409.1) precursor (Met 1-His 232) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4869