Learn More
Sino Biological Recombinant Anti-IL-4R Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10402R102100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL4R (rh IL4R; Catalog#10402-H08H; NP_000409.1; Met 1-His 232).
Catalog No. 50-172-5458