Learn More
Sino Biological Anti-IL-4R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10402RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL4R / CD124 (rh IL4R; Catalog#10402-H08H; NP_000409.1; Met 1-His 232). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5468