Learn More
Sino Biological Anti-GRP78/HSPA5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 104042T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse GRP78/Hspa5, and purified by antigen affinity chromatography.
Catalog No. 50-172-5474