Learn More
Sino Biological Recombinant Human Mannan Binding Lectin/MBL2 Protein, HPLC-verified
Supplier:  Sino Biological 10405HNAS100
Encompass_Preferred
A DNA sequence encoding the human MBL (NP_000233.1) (Met1-Ile 248) was expressed.
Catalog No. 50-161-4872