Learn More
Sino Biological Anti-Kallikrein 1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10407MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human KLK1 / Kallikrein 1 (rh KLK1 / Kallikrein 1; Catalog#10407-H08H; NP_002248.1; Met 1-Ser 262). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5477