Learn More
Sino Biological Anti-Kallikrein 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10407RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KLK1 (rh KLK-1; Catalog#10407-H08H; NP_002248.1; Met 1-Ser 262). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5482