Learn More
Sino Biological Recombinant Human DR4/TRAIL R1 Protein (hFc Tag)
Catalog No. 501614876
Encompass_Preferred
A DNA sequence encoding the human TNFRSF10A (NP_003835.2) extracellular domain (Met 1-Asn 239) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4876 Supplier Sino Biological Supplier No. 10408H02H100
Add to Cart
Add to Cart
A DNA sequence encoding the human TNFRSF10A (NP_003835.2) extracellular domain (Met 1-Asn 239) was fused with the Fc region of human IgG1 at the C-terminus.