Learn More
Sino Biological Recombinant Human DR4/TRAIL R1 Protein (His Tag), HPLC-verified
Catalog No. 501614880
Encompass_Preferred
A DNA sequence encoding the human TNFRSF10A (NP_003835.2) extracellular domain (Met 1-Asn 239) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4880 Supplier Sino Biological Supplier No. 10408H08H100
Add to Cart
Add to Cart
A DNA sequence encoding the human TNFRSF10A (NP_003835.2) extracellular domain (Met 1-Asn 239) was expressed, fused with a polyhistidine tag at the C-terminus.