Learn More
Sino Biological Recombinant Anti-DR4/TRAIL R1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10408R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAIL R1 / CD261 / TNFRSF10A (rh TRAIL R1 / CD261 / TNFRSF10A; Catalog#10408-H08H; NP_003835.2; Met1-Asn239).
Catalog No. 50-172-5503