Learn More
Sino Biological Anti-TRAIL/TNFSF10 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10409MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFSF10 / TRAIL / APO-2L (CD253) (rh TNFSF10 / TRAIL / APO-2L (CD253); Catalog#10409-HNAE; NP_003801.1; Val114-Gly281). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5517