Learn More
Sino Biological Recombinant Human Hemojuvelin Protein (His Tag)
Supplier:  Sino Biological 10410H08B100
Encompass_Preferred
A DNA sequence encoding the mature form of human HFE2 isoform a (Q6ZVN8-1) (Met 1-Ser 399) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4885