Learn More
Sino Biological Recombinant Human DcR2/TRAIL R4 Protein (His Tag)
Catalog No. 501614889
Encompass_Preferred
A DNA sequence encoding the human TNFRSF10D (NP_003831.2) extracellular domain (Met 1-His 211) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4889 Supplier Sino Biological Supplier No. 10413H08H100
A DNA sequence encoding the human TNFRSF10D (NP_003831.2) extracellular domain (Met 1-His 211) was fused with a polyhistidine tag at the C-terminus.