Learn More
Sino Biological Anti-DcR2/TRAIL R4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10413MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TRAIL R4 / CD264 / TNFRSF10D (rh TRAIL R4 / CD264 / TNFRSF10D; Catalog#10413-H08H; NP_003831.2; Met1-His211). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5540