Learn More
Sino Biological Recombinant Anti-DcR2/TRAIL R4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10413R009100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAIL R4 / CD264 / TNFRSF10D (rh TRAIL R4 / CD264 / TNFRSF10D; Catalog#10413-H08H; NP_003831.2; Met1-His211).
Catalog No. 50-172-5543