Learn More
Sino Biological Anti-CD89/FCAR Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10414MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD89 / FCAR (rh CD89; Catalog#10414-H08H; NP_001991.1; Met 1-Asn 227). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5563