Learn More
Sino Biological Recombinant Anti-DcR1/TRAILR3 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10415R003F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAILR3/TNFRSF10C (rh TRAILR3/TNFRSF10C; Catalog#10415-H08H; NP_003832.2; Met1-Pro235) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-5590