Learn More
Sino Biological Recombinant Human TNFR-2 / CD120b Protein (ECD, His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 10417H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Asp257) of human TNFRII (NP_001057.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4897