Learn More
Sino Biological Anti-TNFR-2 / CD120b Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10417MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFR2 / CD120b / TNFRSF1B (rh TNFR2 / CD120b / TNFRSF1B; Catalog#10417-H08H; NP_001057.1; Met1-Asp257). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5600