Learn More
Sino Biological Recombinant Human MKK6 Protein
Supplier:  Sino Biological 10422HNCB50
Encompass_Preferred
A DNA sequence encoding the human MKK6 (NP_002749.2) (Met 1-Asp334) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-4910