Learn More
Sino Biological Anti-Vitronectin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10424MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Vitronectin / VTN (rh Vitronectin / VTN; Catalog#10424-H07H; NP_000629.3; Asp20-Leu478). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5636