Learn More
Sino Biological Anti-Vitronectin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10424RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Vitronectin (rh VTN; Catalog#10424-H07H; NP_000629.3; Asp20-Leu478). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5647