Learn More
Sino Biological Recombinant Human, Mouse, Rat, Rhesus, Canine BMP-2 Protein
Catalog No. 501614916
Encompass_Preferred
A DNA sequence encoding the mature form of human / mouse / rat / rhesus / canine BMP2 (NP_001191.1) (Gln283-Arg396) was expressed with an initial Met. The mature form sequences of human, mouse, rat, rhesus and canine BMP2 are identical.
Catalog No. 50-161-4916 Supplier Sino Biological Supplier No. 10426HNAE100
A DNA sequence encoding the mature form of human / mouse / rat / rhesus / canine BMP2 (NP_001191.1) (Gln283-Arg396) was expressed with an initial Met. The mature form sequences of human, mouse, rat, rhesus and canine BMP2 are identical.