Learn More
Sino Biological Recombinant Human CD204/MSR1 Protein (His Tag)
Catalog No. 501614920
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Lys 77-Leu 451) of human scavenger receptor 1 isoform type 1 (NP_619729.1) was fused with the polyhistidine tag at the N-terminus.
Catalog No. 50-161-4920 Supplier Sino Biological Supplier No. 10427H07H100
A DNA sequence encoding the extracellular domain (Lys 77-Leu 451) of human scavenger receptor 1 isoform type 1 (NP_619729.1) was fused with the polyhistidine tag at the N-terminus.