Learn More
Sino Biological Anti-CD204/MSR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10427RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MSR1 / CD204 (rh MSR1; Catalog#10427-H07H; NP_619729.1; Lys77-Leu451). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5681