Learn More
Sino Biological Anti-CD204/MSR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10427T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human MSR1 / CD204 (rh MSR1 / CD204; Catalog#10427-H07H; NP_619729.1; Lys77-Leu451). MSR1 / CD204 specific IgG was purified by Human MSR1 / CD204 affinity chromatography.
Catalog No. 50-172-5684