Learn More
Sino Biological Recombinant Human TROP-2 Protein (His & hFc Tag)
Catalog No. 501614922
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 274) of human TROP2 (NP_002344.2) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4922 Supplier Sino Biological Supplier No. 10428H03H100
A DNA sequence encoding the extracellular domain (Met 1-Thr 274) of human TROP2 (NP_002344.2) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.