Learn More
Sino Biological Recombinant Human TROP-2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10428H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Thr274) of human TROP2 (NP_002344.2) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4924